2,1,3-Benzothiadiazol-4-ylamine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Aminobenzo-2,1,3-thiadiazole, 4-Aminopiazthiole, Benzo[c][1,2,5]thiadiazol-4-amine, 4-Amino-2,1,3-benzothiadiazole

A151012-1G A151012-5G
ALA151012-1GEA 49.31 USD
ALA151012-1G ALA151012-5G
2,1,3-Benzothiadiazol-4-ylamine ≥98%
2,1,3-Benzothiadiazol-4-ylamine

4-Amino-2,1,3-benzothiadiazole is a gold green to yellow brown powder. It undergoes palladium catalyzed C-N coupling with 2-(2′-bromo-9′,9′-diethylfluoren-7′-yl)-9,9-diethylfluorene to form novel fluorescent dyes. 4-Amino-2,1,3-benzothiadiazole forms dicopper complexes.4-Amino-2,1,3-benzothiadiazole was used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

Formula: C₆H₅N₃S
MW: 151.19 g/mol
Melting Pt: 67-70°C
Storage Temperature: Ambient
MDL Number: MFCD00005810
CAS Number: 767-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2514b27a360c679ee3e9801586d5a731ff42330e&vid=2514b27a360c679ee3e9801586d5a731ff42330e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR