2-Amino-1,3,4-thiadiazole ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

A151011-100G A151011-25G
ALA151011-100GEA 107.26 USD
ALA151011-100G ALA151011-25G
2-Amino-1,3,4-thiadiazole ≥98% (by titration)
2-Amino-1,3,4-thiadiazole
Formula: C₂H₃N₃S
MW: 101.13 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00003107
CAS Number: 4005-51-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=763f6b78c3eff1b628fdce4d1efbc528cae2b7c4&vid=763f6b78c3eff1b628fdce4d1efbc528cae2b7c4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR