9-Acridinecarboxylic acid hydrate ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151008-1G A151008-200MG A151008-25G A151008-5G
ALA151008-1GEA 42.66 USD
ALA151008-1G ALA151008200MG ALA151008-25G ALA151008-5G
9-Acridinecarboxylic acid hydrate ≥97% (by HPLC)
9-Acridinecarboxylic acid hydrate

Product Application:9-Acridinecarboxylic acid hydrate has been used to synthesize a DNA intercalator in order to capture double-stranded DNAs (dsDNAs) duplex from the solution.

Formula: C₁₄H₁₁NO₃
MDL Number: MFCD00149578
CAS Number: 332927-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e7bc4dec1305b106d6ee534981bc8e1ab62650f&vid=6e7bc4dec1305b106d6ee534981bc8e1ab62650f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR