ɑ-Acetamidocinnamic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Acetamido-3-phenylacrylic acid, α-Acetamidocinnamic acid

A151001-100G A151001-25G A151001-5G
ALA151001-100GEA 150.01 USD
ALA151001-100G ALA151001-25G ALA151001-5G
ɑ-Acetamidocinnamic acid ≥98% (by HPLC)
alpha-Acetamidocinnamic acid

Asymmeteric hydrogenation of α-acetamidocinnamic acid with chiral rhodium (I) complexes on charcoal has been reported. α-Acetamidocinnamic acid undergoes asymmetric hydrogenation in ionic liquid catalyzed by rhodium complex to yield (S)-phenylalanine.

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
Melting Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00188416
CAS Number: 5469-45-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e3ffa38757591a3a82fea70f2d9cf935c84d754&vid=2e3ffa38757591a3a82fea70f2d9cf935c84d754&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR