Ammonium Paratungstat ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

A141185-100G A141185-25G
ALA141185-100GEA 39.81 USD
ALA141185-100G ALA141185-25G
Ammonium Paratungstat ≥99%
Ammonium Paratungstat

Ammonium tungsten oxide hydrate is used in production of tungsten alloys. It is used, refluxing the ammonia solution is advisable to speed up the dissolution of the oxide.

Formula: H₄₂N₁₀O₄₂W₁₂ · xH₂O
Melting Pt: >300 °C
Density: 2.3 g/cm³
MDL Number: MFCD00015956
CAS Number: 11120-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fead5cefce6123f8f01851c4b818e33c81b1e0a&vid=7fead5cefce6123f8f01851c4b818e33c81b1e0a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR