Anthrone ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Anthracen-9(10H)-one

A140968-100G A140968-25G
ALA140968-100GEA 208.91 USD
ALA140968-100G ALA140968-25G
Anthrone ≥96%
Anthrone

Anthrone is a trycyclic aromatic hydrocarbon that can be used to determine hydrocarbons using a colorimetric method. It can help estimate carbohydrates in yeast and plant extracts.An aromatic hydrocarbon used to determine hydrocarbons.

Formula: C₁₄H₁₀O
MW: 194.23 g/mol
Boiling Pt: 721°C
Melting Pt: 155°C
Density: 1.194 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3a718401c446353925f1f5f8e5797a7e53747da&vid=b3a718401c446353925f1f5f8e5797a7e53747da&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR