Amoxicillin trihydrate ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
6-[2-Amino-2-(4-hydroxyphenyl)-acetyl]amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, Amoxicillin Ttrihydrate, (2S,5R,6S)-6-((S)-2-amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
A140484-100G
A140484-25G
A140484-5G
A140484-1G
ALA140484-100GEA
377.06
USD
ALA140484-100G
ALA140484-25G
ALA140484-5G
ALA140484-1G
Amoxicillin trihydrate ≥98%
Amoxicillin trihydrate
Formula:
C₁₆H₁₉N₃O₅S MW: 365.40 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00072029 CAS Number: 61336-70-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...