Ammonium metatungstate ≥99.5% (metals basis)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Ammonium tungsten oxide hydrate, AMT, Ammonium tungsten partial, Ammonium metatungstate hydrate

A140379-25G A140379-500G A140379-100G
ALA140379-25GEA 41.71 USD
ALA140379-25G ALA140379-500G ALA140379-100G
Ammonium metatungstate ≥99.5% (metals basis)
Ammonium metatungstate
Formula: (NH₄)₆H₂W₁₂O₄₀·XH₂O
MW: 2956.26 g/mol (anhydrous basis)
Melting Pt: 100°C
Density: 2.3 g/cm³
MDL Number: MFCD00150662
CAS Number: 12333-11-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc03425d51c338e078c8810ef792bce8f19032a5&vid=fc03425d51c338e078c8810ef792bce8f19032a5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR