(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), 7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, (7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate, (7-Azabenzotriazol-1-yl)-N-oxy-tris(dimethyamino)phosphonium hexafluorosphate, AOP, Tris(dimethylamino)(3H-1,2,3-triazolo[4,5-b]pyridin-3-yloxy)phosphonium Hexafluorophosphate

A140263-1G A140263-5G
ALA140263-1GEA 23.66 USD
ALA140263-1G ALA140263-5G
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) ≥96%
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )
Formula: C₁₂H₂₂F₆N₆OP₂
MW: 443.27 g/mol
Melting Pt: 177 °C
MDL Number: MFCD09263289
CAS Number: 156311-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e50e39a3f72d0eeae47ae6151d63ebc0ae5a2bc7&vid=e50e39a3f72d0eeae47ae6151d63ebc0ae5a2bc7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR