N-Fmoc-8-aminooctanoic acid ≥98% (by HPLC)

Supplier: Aladdin Scientific

A140203-5G A140203-10G A140203-1G A140203-250MG
ALA140203-5GEA 318.16 USD
ALA140203-5G ALA140203-10G ALA140203-1G ALA140203250MG
N-Fmoc-8-aminooctanoic acid ≥98% (by HPLC)
N-Fmoc-8-aminooctanoic acid
Formula: C₂₃H₂₇NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00235890
CAS Number: 126631-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f1fb7d53bf8c673421473edf3c1a14cf2b255a9&vid=8f1fb7d53bf8c673421473edf3c1a14cf2b255a9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR