2-Amino-5-methylbenzonitrile ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A140195-1G A140195-250MG A140195-25G A140195-5G
ALA140195-1GEA 51.21 USD
ALA140195-1G ALA140195250MG ALA140195-25G ALA140195-5G
2-Amino-5-methylbenzonitrile ≥97% (by GC)
2-Amino-5-methylbenzonitrile
Formula: C₈H₈N₂
Boiling Pt: 152 °C
Melting Pt: 59-63°C
Storage Temperature: Refrigerator
MDL Number: MFCD00267506
CAS Number: 5925-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8500093e65867185094afce1b628c11f3e5ad63e&vid=8500093e65867185094afce1b628c11f3e5ad63e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR