2-Amino-5-bromo-3-(hydroxymethyl)pyridine ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
2-Amino-5-bromo-3-pyridinemethanol
A140189-1G
A140189-250MG
ALA140189-1GEA
43.61
USD
ALA140189-1G
ALA140189250MG
2-Amino-5-bromo-3-(hydroxymethyl)pyridine ≥98%
2-Amino-5-bromo-3-(hydroxymethyl)pyridine
Formula:
C₆H₇BrN₂O MW: 203.039 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05664025 CAS Number: 335031-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...