4-(4-Aminophenyl)morpholin-3-one ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-(3-Oxomorpholino)aniline

A140078-5G A140078-100G A140078-25G
ALA140078-5GEA 36.96 USD
ALA140078-5G ALA140078-100G ALA140078-25G
4-(4-Aminophenyl)morpholin-3-one ≥98%
4-(4-Aminophenyl)morpholin-3-one
Formula: C₁₀H₁₂N₂O₂
MW: 192.2145 g/mol
Melting Pt: 171.0-175.0°C
Storage Temperature: Ambient
MDL Number: MFCD08236742
CAS Number: 438056-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80f9a3e8e2984ce0e106c1b53dc4ebf1b6f43c15&vid=80f9a3e8e2984ce0e106c1b53dc4ebf1b6f43c15&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR