Aniline-2,5-disulfonic acid monosodium salt ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Sodium Hydrogen 2-Amino-1,4-benzenedisulphonate, 2,5-Disulfoaniline Monosodium Salt, Aniline-2,5-disulphonic Acid Monosodium Salt, 2,5-Disulphoaniline Monosodium Salt, Sodium Hydrogen 2-Amino-1,4-benzenedisulfonate, 2-Amino-1,4-benzenedisulfonic acid Monosodium Salt, Sodium 2-amino-4-sulphobenzenesulphonate, 2-Amino-1,4-benzenedisulphonic acid Monosodium Salt

A139896-100G A139896-25G
ALA139896-100GEA 133.86 USD
ALA139896-100G ALA139896-25G
Aniline-2,5-disulfonic acid monosodium salt ≥98% (by HPLC)
Aniline-2,5-disulphonic acid monosodium salt
Formula: C₆H₇NO₆S₂
MW: 275.22 g/mol
MDL Number: MFCD00078393
CAS Number: 24605-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b16f181cf1c7a81f6010a170532b19788750437&vid=7b16f181cf1c7a81f6010a170532b19788750437&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR