6-Amino-2-naphthalenesulfonic Acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A139448-100G A139448-250G A139448-25G
ALA139448-100GEA 175.66 USD
ALA139448-100G ALA139448-250G ALA139448-25G
6-Amino-2-naphthalenesulfonic Acid ≥98% (by HPLC)
6-Amino-2-naphthalenesulfonic Acid

6-Amino-2-naphthalenesulfonic Acid is used in the synthesis of diaryl urea derivatives bearing sulfonamide moiety with an in vitro antitumor activity.

Formula: C₁₀H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00035736
CAS Number: 93-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ba3c9b295618cc4b1458fab95fd8d42d75d722&vid=58ba3c9b295618cc4b1458fab95fd8d42d75d722&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR