2-Amino-3,5-dibromo-6-methylpyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3,5-Dibromo-6-methyl-2-pyridinamine

A139439-100G A139439-1G A139439-25G A139439-5G
ALA139439-100GEA 2651.36 USD
ALA139439-100G ALA139439-1G ALA139439-25G ALA139439-5G
2-Amino-3,5-dibromo-6-methylpyridine ≥98% (by GC)
2-Amino-3,5-dibromo-6-methylpyridine

Product Application:2-Amino-3,5-dibromo-6-methylpyridine may be used to synthesize 1-[[(3,5-dibromo-6-methyl-2-pyridinyl)imino]-methylenyl]-2-naphthalenol and 3,5-dibromo-6-methyl-2-pyridone.

Formula: C₆H₆Br₂N₂
MW: 265.94
Melting Pt: 134°C
MDL Number: MFCD00068229
CAS Number: 91872-10-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebd584a382a31dac5b0e41c46dcfbf91c6b0acf3&vid=ebd584a382a31dac5b0e41c46dcfbf91c6b0acf3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR