5-Bromo-2-methylpyridin-3-amine ≥96%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
3-Amino-5-bromo-2-methylpyridine
A139435-1G
A139435-250MG
A139435-25G
A139435-5G
ALA139435-1GEA
206.06
USD
ALA139435-1G
ALA139435250MG
ALA139435-25G
ALA139435-5G
5-Bromo-2-methylpyridin-3-amine ≥96%
5-Bromo-2-methylpyridin-3-amine
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol Melting Pt: 108-109°C Storage Temperature: Ambient |
MDL Number:
MFCD09031418 CAS Number: 914358-73-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...