3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester, 2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester

A139401-1G A139401-250MG A139401-5G
ALA139401-1GEA 269.71 USD
ALA139401-1G ALA139401250MG ALA139401-5G
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥96%
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester

Application:It is used as a pharmaceutical intermediate.

Formula: C₁₂H₁₉BN₂O₃
Melting Pt: 144 °C
MDL Number: MFCD12923427
CAS Number: 893440-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daea1bb1caad010a9e39c8636972facbcf8aa5de&vid=daea1bb1caad010a9e39c8636972facbcf8aa5de&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR