6-Amidino-2-naphthol Methanesulfonate ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A139303-1G A139303-250MG A139303-25G A139303-5G
ALA139303-1GEA 111.06 USD
ALA139303-1G ALA139303250MG ALA139303-25G ALA139303-5G
6-Amidino-2-naphthol Methanesulfonate ≥98% (by HPLC)
6-Amidino-2-naphthol Methanesulfonate
Formula: C₁₁H₁₀N₂O
MDL Number: MFCD00143493
CAS Number: 82957-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c86250b86faddb0c1ca54d40167dca4077ac006d&vid=c86250b86faddb0c1ca54d40167dca4077ac006d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR