2-Acetylamino-4-methylthiazole-5-sulfonyl chloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Acetylamino-4-methyl-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-1,3-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methylthiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-5-thiazolesulphonyl chloride

A139197-1G A139197-250MG
ALA139197-1GEA 54.06 USD
ALA139197-1G ALA139197250MG
2-Acetylamino-4-methylthiazole-5-sulfonyl chloride ≥97%
2-Acetylamino-4-methylthiazole-5-sulfonyl chloride

2-Acetamido-4-methyl-5-thiazolesulfonyl chloride was used in high-performance liquid chromatographic (HPLC) assay for acetazolamide1.

Formula: C₆H₇ClN₂O₃S₂
Melting Pt: 158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005320
CAS Number: 69812-29-9
EINECS: 274-127-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e20417fa85cff62c16198474ee749c0c0bf82c82&vid=e20417fa85cff62c16198474ee749c0c0bf82c82&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR