4-(Aminomethyl)phenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Hydroxybenzylamine

A139192-100G A139192-25G A139192-5G
ALA139192-100GEA 182.31 USD
ALA139192-100G ALA139192-25G ALA139192-5G
4-(Aminomethyl)phenol ≥98%
4-(Aminomethyl)phenol
Formula: C₇H₉NO
MW: 123.16 g/mol
Melting Pt: 115°C
Storage Temperature: Ambient
MDL Number: MFCD19690971
CAS Number: 696-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d01d101cd78d3eb3c65987b3eb07094ecfc6a846&vid=d01d101cd78d3eb3c65987b3eb07094ecfc6a846&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR