8-O-Acetylharpagide ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 8-Acetylharpagide, (1S,4aS,5R,7S,7aS)-4a,5-Dihydroxy-7-methyl-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-7-yl acetate, Harpagide 8-acetate, 8-O-Acetylharpagide

A139182-25MG A139182-5MG A139182-100MG A139182-1MG
ALA139182-25MGEA 151.91 USD
ALA139182-25MG ALA139182-5MG ALA139182100MG ALA139182-1MG
8-O-Acetylharpagide ≥98%
8-O-Acetylharpagide
Formula: C₁₇H₂₆O₁₁
MW: 406.38 g/mol
Melting Pt: 225 - 228 °C
Storage Temperature: Ambient
MDL Number: MFCD03700750
CAS Number: 6926-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05091f4de68379b44aa5cea72df0d2948c756450&vid=05091f4de68379b44aa5cea72df0d2948c756450&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR