6-Amino-1,3-dimethyluracil ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 6-Amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 4-Amino-1,3-Dimethyluracil

A139156-100G A139156-25G
ALA139156-100GEA 97.76 USD
ALA139156-100G ALA139156-25G
6-Amino-1,3-dimethyluracil ≥98%
6-Amino-1,3-dimethyluracil
Formula: C₆H₉N₃O₂
MW: 155.1546 g/mol
MDL Number: MFCD00006552
CAS Number: 6642-31-5
EINECS: 229-662-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=003edadfaae1fcbb8bc82c51d1524a8065795723&vid=003edadfaae1fcbb8bc82c51d1524a8065795723&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR