2-Amino-6-chloro-4-nitrophenol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A139127-1G A139127-25G A139127-5G
ALA139127-1GEA 18.91 USD
ALA139127-1G ALA139127-25G ALA139127-5G
2-Amino-6-chloro-4-nitrophenol ≥97%
2-Amino-6-chloro-4-nitrophenol
Formula: C₆H₅ClN₂O₃
Melting Pt: 162 °C
MDL Number: MFCD00035767
CAS Number: 6358-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR