5-Amino-2-chlorophenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A139126-100G A139126-10G A139126-1G A139126-25G A139126-5G
ALA139126-100GEA 539.51 USD
ALA139126-100G ALA139126-10G ALA139126-1G ALA139126-25G ALA139126-5G
5-Amino-2-chlorophenol ≥98%
5-Amino-2-chlorophenol
Formula: C₆H₆ClNO
Melting Pt: 160°C
Storage Temperature: Ambient
MDL Number: MFCD01707858
CAS Number: 6358-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR