4-Acetamidopiperidine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A139032-5G A139032-1G
ALA139032-5GEA 109.16 USD
ALA139032-5G ALA139032-1G
4-Acetamidopiperidine ≥98% (by GC)
4-Acetamidopiperidine
Formula: C₇H₁₄N₂O
Melting Pt: 141°C
MDL Number: MFCD05254805
CAS Number: 5810-56-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9569c4c83b97ce66c02716cef47508e6f38cf5ee&vid=9569c4c83b97ce66c02716cef47508e6f38cf5ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR