3-Acetyl-2-chloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-acetyl-2-chloropyridine, 1-(2-Chloro-3-pyridinyl)-1-ethanone, 1-(2-Chloropyridin-3-yl)ethanone

A138997-1G A138997-5G
ALA138997-1GEA 63.56 USD
ALA138997-1G ALA138997-5G
3-Acetyl-2-chloropyridine ≥98%
3-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
Boiling Pt: 114 °C
Density: 1.27 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD03840751
CAS Number: 55676-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7aaf45331b5b79745bec20d624338e3514e38f21&vid=7aaf45331b5b79745bec20d624338e3514e38f21&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR