5-Bromo-1,3,4-thiadiazol-2-amine ≥95% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A138762-1G
A138762-25G
A138762-5G
ALA138762-1GEA
35.06
USD
ALA138762-1G
ALA138762-25G
ALA138762-5G
5-Bromo-1,3,4-thiadiazol-2-amine ≥95% (by HPLC)
5-Bromo-1,3,4-thiadiazol-2-amine
Formula:
C₂H₂BrN₃S |
MDL Number:
MFCD00464325 CAS Number: 37566-39-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...