4-Amino-2,6-dimethylpyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A138719-1G A138719-250MG A138719-25G A138719-5G
ALA138719-1GEA 76.86 USD
ALA138719-1G ALA138719250MG ALA138719-25G ALA138719-5G
4-Amino-2,6-dimethylpyridine ≥98%
4-Amino-2,6-dimethylpyridine

Application:4-Amino-2,6-dimethylpyridine, is an important raw material and intermediate used in pharmaceuticals, agrochemicals, dyestuff, field.

Formula: C₇H₁₀N₂
Boiling Pt: 246°C
Melting Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00130078
CAS Number: 3512-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68fcaedad45003d45fc8de494a1eba6ac14d87cd&vid=68fcaedad45003d45fc8de494a1eba6ac14d87cd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR