3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-Amino-3-(3,4-dimethoxyphenyl)propionic acid

A138705-1G A138705-25G A138705-5G
ALA138705-1GEA 32.21 USD
ALA138705-1G ALA138705-25G ALA138705-5G
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid ≥98% (by HPLC, titration analysis)
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid
Formula: C₁₁H₁₅NO₄
MW: 225.244 g/mol
Melting Pt: 223°C
MDL Number: MFCD00735172
CAS Number: 34841-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b20ded359d451318ff162c744924c4cd279eeae&vid=7b20ded359d451318ff162c744924c4cd279eeae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR