3-Amino-3-(4-fluorophenyl)propionic acid ≥98% (by titration)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A138673-1G
A138673-250MG
ALA138673-1GEA
45.51
USD
ALA138673-1G
ALA138673250MG
3-Amino-3-(4-fluorophenyl)propionic acid ≥98% (by titration)
3-Amino-3-(4-fluorophenyl)propionic acid
Formula:
C₉H₁₀FNO₂ MW: 183.18 g/mol Melting Pt: 216°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00181811 CAS Number: 325-89-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...