ω-Acetoxyacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A138520-100G A138520-25G
ALA138520-100GEA 208.91 USD
ALA138520-100G ALA138520-25G
ω-Acetoxyacetophenone ≥98%
ω-Acetoxyacetophenone
Formula: C₆H₅C(O)CH₂OC(O)CH₃
MW: 178.19 g/mol
Boiling Pt: 110-114°C
Melting Pt: 48-50°C
Flash Pt: >110°C
MDL Number: MFCD00026205
CAS Number: 2243-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9df3030301e6c0d91217a189f05ffd81f55e816&vid=d9df3030301e6c0d91217a189f05ffd81f55e816&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR