2-Amino-5-methylthiophene-3-carbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-5-methyl-3-thiophenecarbonitrile, 2-Amino-3-cyano-5-methylthiophene

A138196-1G A138196-25G A138196-5G
ALA138196-1GEA 9.41 USD
ALA138196-1G ALA138196-25G ALA138196-5G
2-Amino-5-methylthiophene-3-carbonitrile ≥98%
2-Amino-5-methylthiophene-3-carbonitrile
Formula: C₆H₆N₂S
MW: 138.1902 g/mol
Melting Pt: 103°C
Storage Temperature: Ambient
MDL Number: MFCD01626055
CAS Number: 138564-58-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=109d6bbc1c970a4400098fbdd859b939bce8d6f4&vid=109d6bbc1c970a4400098fbdd859b939bce8d6f4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR