4-Amino-3-furazanecarboxamidoxime ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A138171-250MG A138171-5G A138171-1G
ALA138171250MGEA 14.16 USD
ALA138171250MG ALA138171-5G ALA138171-1G
4-Amino-3-furazanecarboxamidoxime ≥98%
4-Amino-3-furazanecarboxamidoxime
Formula: C₃H₅N₅O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00186407
CAS Number: 13490-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d482552002c99f251ecab7bde82b93db02182e74&vid=d482552002c99f251ecab7bde82b93db02182e74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR