2-Amino-4'-bromobenzophenone ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A138046-1G A138046-5G
ALA138046-1GEA 62.61 USD
ALA138046-1G ALA138046-5G
2-Amino-4'-bromobenzophenone ≥95%
2-Amino-4'-bromobenzophenone
Formula: C₁₃H₁₀BrNO
Melting Pt: 111 °C
MDL Number: MFCD02181034
CAS Number: 1140-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29850a0eedffabd823cd728d66f8328fbeb7a63b&vid=29850a0eedffabd823cd728d66f8328fbeb7a63b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR