1,3-benzothiazol-5-amine ≥96%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole
A138033-100G
A138033-25G
A138033-5G
ALA138033-100GEA
3523.46
USD
ALA138033-100G
ALA138033-25G
ALA138033-5G
1,3-benzothiazol-5-amine ≥96%
1,3-benzothiazol-5-amine
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Formula:
C₇H₆N₂S MW: 150.199 g/mol Melting Pt: 55-59°C |
MDL Number:
MFCD04115282 CAS Number: 1123-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...