4-Amino-5-chloro-2-ethoxybenzoic acid ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
4-Amino-5-chloro-2-ethoxybenzoic Acid
A138001-25G
A138001-5G
ALA138001-25GEA
66.41
USD
ALA138001-25G
ALA138001-5G
4-Amino-5-chloro-2-ethoxybenzoic acid ≥98% (by HPLC)
4-Amino-5-chloro-2-ethoxybenzoic acid
Formula:
C₉H₁₀ClNO₃ MW: 215.63 g/mol Melting Pt: 169 °C |
MDL Number:
MFCD03840527 CAS Number: 108282-38-8 EINECS: 600-861-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...