L(+)-Arginine ≥99% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

A137768-100G A137768-25G A137768-500G
ALA137768-100GEA 17.01 USD
ALA137768-100G ALA137768-25G ALA137768-500G
L(+)-Arginine ≥99% (by HPLC), Moligand™
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 368
Melting Pt: 238
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ba3c032682f84b32f781618f7f1fb0abea4577f&vid=9ba3c032682f84b32f781618f7f1fb0abea4577f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR