3-Aminobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A137766-100G A137766-250G A137766-25G
ALA137766-100GEA 32.21 USD
ALA137766-100G ALA137766-250G ALA137766-25G
3-Aminobenzoic acid ≥98%
3-Aminobenzoic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 174°C
Density: 1.51 g/cm³
Flash Pt: 150 °C
MDL Number: MFCD00007795
CAS Number: 99-05-8
EINECS: 202-724-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR