Amoxicillin sodium ≥89%

Supplier: ALADDIN SCIENTIFIC
Danger

A137746-25G A137746-5G A137746-100G A137746-1G
ALA137746-25GEA 327.66 USD
ALA137746-25G ALA137746-5G ALA137746-100G ALA137746-1G
Amoxicillin sodium ≥89%
Amoxicillin sodium

Amoxicillin Sodium is a moderate- spectrum, bacteriolytic, β-lactam antibiotic.

Formula: C₁₆H₁₈N₃NaO₅S
MDL Number: MFCD08063910
CAS Number: 34642-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0540f2712d99c2295b32488ee4a6f2b2c6252f15&vid=0540f2712d99c2295b32488ee4a6f2b2c6252f15&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR