2,6-Dichlorobenzal chloride ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A137698-25G A137698-5G
ALA137698-25GEA 85.41 USD
ALA137698-25G ALA137698-5G
2,6-Dichlorobenzal chloride ≥98% (by GC)
2,6-Dichlorobenzal chloride

It is a halogenated hydrocarbon.

Formula: C₇H₄Cl₄
MDL Number: MFCD00016349
CAS Number: 81-19-6
UN: 3265
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fceabe039f540497e2a0f9410fb71922b09cf335&vid=fceabe039f540497e2a0f9410fb71922b09cf335&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR