2-Aminophenylboronic acid ≥96% (contains varying amounts of Anhydride)

Supplier: ALADDIN SCIENTIFIC
Warning

A137630-100MG A137630-1G A137630-250MG A137630-25G A137630-5G
ALA137630100MGEA 31.26 USD
ALA137630100MG ALA137630-1G ALA137630250MG ALA137630-25G ALA137630-5G
2-Aminophenylboronic acid ≥96% (contains varying amounts of Anhydride)
2-Aminophenylboronic acid
Formula: C₆H₈BNO₂
Melting Pt: 188-190°
Storage Temperature: Ambient
MDL Number: MFCD01074645
CAS Number: 5570-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d18e1356f5a1636e1f90fcc496d3fcf807b0c18&vid=2d18e1356f5a1636e1f90fcc496d3fcf807b0c18&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR