6-Amino-1-methyluracil ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
6-Amino-1-methylpyrimidine-2,4(1H,3H)-dione
Product Description6-Amino-1-methyluracil is an amino derivative of uracil and its standard molar enthalpy of combustion has been evaluated. Host-guest complexation of 6-amino-1-methyluracil with tetrapropoxycalix[4]arene has been reported.Product Application6-Amino-1-methyluracil may be used in the preparation of 1,1′-di methyl-1H-spiro[pyrimido[4,5-b]quinoline-5,5′-pyrrolo[2,3-d]pyrimidine]-2,2′,4,4′,6′(1′H,3H,3′H,7′H,1′H)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid.
Formula:
C₅H₇N₃O₂ Melting Pt: 307 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075366 CAS Number: 2434-53-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...