4-Amino-2-fluoropyridine ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A137501-1G A137501-25G A137501-5G
ALA137501-1GEA 17.96 USD
ALA137501-1G ALA137501-25G ALA137501-5G
4-Amino-2-fluoropyridine ≥96%
4-Amino-2-fluoropyridine
Formula: C₅H₅FN₂
Storage Temperature: Refrigerator
MDL Number: MFCD03095284
CAS Number: 18614-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ee3994c92d0ab35ae398394eea8a0124a77ea9&vid=58ee3994c92d0ab35ae398394eea8a0124a77ea9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR