3-Amino-6-chloro-2-methylpyridine ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Chloro-2-methylpyridin-3-amine

A137489-100G A137489-25G A137489-5G A137489-1G
ALA137489-100GEA 226.01 USD
ALA137489-100G ALA137489-25G ALA137489-5G ALA137489-1G
3-Amino-6-chloro-2-methylpyridine ≥95%
3-Amino-6-chloro-2-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD03095220
CAS Number: 164666-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b0f3276aabeca0087d842c0820fe5cffb69ded&vid=c2b0f3276aabeca0087d842c0820fe5cffb69ded&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR