6-Amino-2-(methylthio)pyrimidin-4-ol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-2-(methylthio)-6-pyrimidinol

A137424-1G A137424-250MG
ALA137424-1GEA 17.96 USD
ALA137424-1G ALA137424250MG
6-Amino-2-(methylthio)pyrimidin-4-ol ≥97%
6-Amino-2-(methylthio)pyrimidin-4-ol
Formula: C₅H₇N₃OS
MW: 157.194 g/mol
MDL Number: MFCD00023240
CAS Number: 1074-41-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ce0df364939462d992e84a34522a092aab83f1b&vid=9ce0df364939462d992e84a34522a092aab83f1b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR