3-Amino-4-fluorophenol ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A137308-5G
A137308-1G
A137308-250MG
ALA137308-5GEA
750.41
USD
ALA137308-5G
ALA137308-1G
ALA137308250MG
3-Amino-4-fluorophenol ≥98%
3-Amino-4-fluorophenol
Formula:
C₆H₆FNO MW: 127.1200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09033850 CAS Number: 62257-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...