ɑ-Cyclopentylphenylacetic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: ɑ-Phenylcyclopentaneacetic Acid, α-Phenylcyclopentaneacetic acid, 2-Cyclopentyl-2-phenylacetic acid, α-Cyclopentylphenylacetic acid

A137219-10G A137219-1G A137219-25G A137219-5G
ALA137219-10GEA 81.61 USD
ALA137219-10G ALA137219-1G ALA137219-25G ALA137219-5G
ɑ-Cyclopentylphenylacetic acid ≥97%
alpha-Cyclopentylphenylacetic acid

Cyclopentylphenylacetic acid was used in the synthesis of: · 1-cyclopentyl-l-phenyl-2-(p-alkoxyphenyl)ethylenes. · soft ester analogs of anticholinergics.

Formula: C₁₃H₁₆O₂
Melting Pt: 100 °C
MDL Number: MFCD00001379
CAS Number: 3900-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1456bc4ca9b9913471259fea1d0ceaa7938b02a&vid=e1456bc4ca9b9913471259fea1d0ceaa7938b02a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR