5-Amino-2-chloro-3-methylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Amino-2-chloro-3-picoline, 6-Chloro-5-methylpyridin-3-amine

A137214-100G A137214-1G A137214-25G A137214-5G
ALA137214-100GEA 1228.26 USD
ALA137214-100G ALA137214-1G ALA137214-25G ALA137214-5G
5-Amino-2-chloro-3-methylpyridine ≥97%
5-Amino-2-chloro-3-methylpyridine
Formula: C₆H₇ClN₂
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD03095087
CAS Number: 38186-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03fa4a0396b9b28be2b5c77671f910f0b3912899&vid=03fa4a0396b9b28be2b5c77671f910f0b3912899&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR