1-Amino-2-methylpropan-2-ol ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 1-Amino-2-methyl-2-propanol, 1,1-Dimethylethanolamine

A137158-100G A137158-1G A137158-25G A137158-5G
ALA137158-100GEA 3869.26 USD
ALA137158-100G ALA137158-1G ALA137158-25G ALA137158-5G
1-Amino-2-methylpropan-2-ol ≥95%
1-Amino-2-methylpropan-2-ol
Formula: C₄H₁₁NO
MW: 89.1362 g/mol
Boiling Pt: 151°C
Density: 0.957 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01697283
CAS Number: 2854-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=defc223e72ded12dcd0e8804a668c6e0b14afd96&vid=defc223e72ded12dcd0e8804a668c6e0b14afd96&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR